missing translation for 'onlineSavingsMsg'
Learn More

L-O-Phosphoserine-13C3, TRC

Product Code. 30425573
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
5 mg
Unit Size:
1mg
2.50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30425573 5 mg 5mg
30413580 2.5 mg 2.50mg
30422541 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30425573 Supplier Toronto Research Chemicals Supplier No. TRCP3612035MG
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material L-O-Phosphoserine-13C3
Molecular Formula 13C3 H8 N O6 P
Quantity 5 mg
Synonym Dihydrogen Phosphate L-Serine-13C3, L-Serine Phosphate-13C3, L-Serine Dihydrogen Phosphate-13C3, 3-O-Phosphoserine-13C3, Dexfosfoserine-13C3, Fosforina-13C3, L-3-Phosphoserine-13C3, L-O-Phosphoserine-13C3, L-O-Serine Phosphate-13C3, L-Phosphoserine-13C3, L-Serinephosphoric Acid-13C3, L-Seryl Phosphate-13C3, O-Phospho-L-serine-13C3, O-Phosphoryl-L-serine-13C3, O-Phosphorylserine-13C3, Phospho-L-serine-13C3, Serine O-Phosphate-13C3, Serine Dihydrogen Phosphate-13C3, Seriphos-13C3
InChI Formula InChI=1S/C3H8NO6P/c4-2(3(5)6)1-10-11(7,8)9/h2H,1,4H2,(H,5,6)(H2,7,8,9)/t2-/m0/s1/i1+1,2+1,3+1
SMILES N[13C@@H]([13CH2]OP(=O)(O)O)[13C](=O)O
IUPAC Name (2S)-2-amino-3-phosphonooxy(1,2,3-13C3)propanoic acid
Molecular Weight (g/mol) 188.05
Formula Weight 188.019
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.