missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ L Moses dihydrochloride

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
L Moses dihydrochloride is a high affinity and selective cell-permeable p300/CBP-associated factor (PCAF) inhibitor (Ki = 47 nM), which exhibits no significant activity against a panel of 48 other bromodomains except GCN5 (Kd = 600 nM). L Moses exhibits >4500-fold selectivity for PCAF over BRD4. The compound inhibits Tunicamycin (Cat. No. 3516) -induced neuronal cell death by reversing the transcriptional changes associated with Tunicamycin treatment. L Moses exhibits metabolic stability in mouse and human liver microsomes and no observable cytotoxicity in peripheral blood mononuclear cells (PBMC).
Specifications
Specifications
| Chemical Name or Material | (1S, 2S)-N1,N1-Dimethyl-N2-(3-methyl-1,2,4-triazolo[3,4-a]phthalazin-6-yl)-1-phenylpropane-1,2-diamine dihydrochloride |
| Assay Percent Range | >98% |
| Quantity | 10 mg |
| Molecular Formula | C21H24N6.2HCl |
| Formula Weight | Observed MW: 433.38 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction