Learn More
L-Lysine-4,4,5,5-d4 Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | L-Lysine-4,4,5,5-d4 Hydrochloride |
| CAS | 284664-96-6 |
| Molecular Formula | C6 2H4 H10 N2 O2 . Cl H |
| Quantity | 25 mg |
| Synonym | L-Lysine-4,4,5,5-d4, hydrochloride (1:1) (ACI), L-Lysine-4,4,5,5-d4, monohydrochloride (9CI), L-Lysine-4,4,5,5-D4 hydrochloride, L-Lysine-4,4,5,5-d4 hydrochloride, L-Lysine-d4 hydrochloride |
| InChI Formula | InChI=1S/C6H14N2O2.ClH/c7-4-2-1-3-5(8)6(9)10;/h5H,1-4,7-8H2,(H,9,10);1H/t5-;/m0./s1/i1D2,2D2; |
| SMILES | Cl.[2H]C([2H])(CN)C([2H])([2H])C[C@H](N)C(=O)O |
| IUPAC Name | (2S)-2,6-diamino-4,4,5,5-tetradeuteriohexanoic acid;hydrochloride |
| Molecular Weight (g/mol) | 186.67 |
| Formula Weight | 186.1073 |