Learn More
L-Lactic Acid-d3 Sodium Salt, TRC
Product Code. 30478893
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30478893 25 mg 25mg
30479919 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30478893 Supplier Toronto Research Chemicals Supplier No. TRCL11350325MG
Please to purchase this item. Need a web account? Register with us today!
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 285979-84-2
Molecular Formula C3 2H3 H2 O3 . Na
Molecular Weight (g/mol) 115.08
Synonym Propanoic-3,3,3-d3 acid, 2-hydroxy-, monosodium salt, (2S)- (9CI), L-(+)-Lactic acid-d3 sodium salt, Sodium S-lactate-d3, Sodium (2S)-2-hydroxypropanoate-3,3,3-d3, (S)-2-Hydroxypropionic-3,3,3-d3 acid sodium salt, Sodium (S)-lactate-3,3,3-d3, Sodium L-(+)-lactate-d3, Sodium L-lactate-3,3,3-D3
IUPAC Name sodium;(2S)-3,3,3-trideuterio-2-hydroxypropanoate
SMILES [Na+].[2H]C([2H])([2H])[C@H](O)C(=O)[O-]
Specifications
Quantity 25 mg
InChI Formula InChI=1S/C3H6O3.Na/c1-2(4)3(5)6;/h2,4H,1H3,(H,5,6);/q;+1/p-1/t2-;/m0./s1/i1D3;
Formula Weight 115.0325
Chemical Name or Material L-Lactic Acid-d3 Sodium Salt