missing translation for 'onlineSavingsMsg'
Learn More

L-Hyoscyamine, MedChemExpress

Product Code. 16482376
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16482376 500 mg 500mg
16479086 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16482376 Supplier Medchem Express Supplier No. HYN0471

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress L-Hyoscyamine (Daturine), a natural plant tropane alkaloid, is a potent and competitive muscarinic receptor (MR) antagonist. L-Hyoscyamine is a levo-isomer to Atropine (HY-B1205).

L-Hyoscyamine (Daturine), a natural plant tropane alkaloid, is a potent and competitive muscarinic receptor (MR) antagonist. L-Hyoscyamine is a levo-isomer to Atropine (HY-B1205).

Specifications

Chemical Name or Material L-Hyoscyamine
CAS 101-31-5
Color White
Purity Grade Notes Research
Molecular Formula C17H23NO3
Quantity 500 mg
Synonym Daturine
Solubility Information DMSO : 100 mg/mL (345.58 mM; Need ultrasonic)
SMILES CN1[C@@H]2C[C@@H](OC([C@@H](C3=CC=CC=C3)CO)=O)C[C@H]1CC2
Molecular Weight (g/mol) 289.37
Formula Weight 289.37
Percent Purity 98.2%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.