Learn More
L-Homocysteine Thiolactone Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | L-Homocysteine Thiolactone Hydrochloride |
| CAS | 31828-68-9 |
| Molecular Formula | C4H8ClNOS |
| Quantity | 1 g |
| Synonym | (3S)-3-Aminodihydro-2(3H)-thiophenone Hydrochloride, (S)-3-Aminodihydro-2(3H)-thiophenone Hydrochloride, L-3-Aminodihydro-2(3H)-thiophenone Hydrochloride |
| InChI Formula | InChI=1S/C4H7NOS.ClH/c5-3-1-2-7-4(3)6;/h3H,1-2,5H2;1H/t3-;/m0./s1 |
| SMILES | N[C@@H](CCS1)C1=O.Cl |
| Molecular Weight (g/mol) | 153.63 |
| Formula Weight | 153.0015 |