Learn More
L-Glutamine Methyl Ester di-Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | L-Glutamine Methyl Ester di-Hydrochloride |
| CAS | 32668-14-7 |
| Molecular Formula | C6 H12 N2 O3 . 2[HCl] |
| Quantity | 2.5 g |
| Synonym | L-Glutamine Methyl Ester Monohydrochloride |
| InChI Formula | InChI=1S/C6H12N2O3.2ClH/c1-11-6(10)4(7)2-3-5(8)9;;/h4H,2-3,7H2,1H3,(H2,8,9);2*1H/t4-;;/m0../s1 |
| SMILES | Cl.Cl.COC(=O)[C@@H](N)CCC(=O)N |
| IUPAC Name | methyl (2S)-2,5-diamino-5-oxopentanoate;dihydrochloride |
| Molecular Weight (g/mol) | 233.093 |
| Formula Weight | 232.038 |