Learn More
L-Glutamic Acid-d5, TRC
Specifications
Specifications
| Chemical Name or Material | L-Glutamic Acid-d5 |
| CAS | 2784-50-1 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C5 2H5 H4 N O4 |
| Quantity | 50 mg |
| Synonym | L-Glutamic-2,3,3,4,4-d5 acid (9CI, ACI), Glutamic-2,3,3,4,4-d5 acid, L- (7CI, 8CI), L-[2,3,3,4,4-2H5]Glutamic acid, Glutamic-2,3,3,4,4-d5 acid, L-Glutamic Acid-2,3,3,4,4-D5, (2S)-2-Aminopentanedioic-2,3,3,4,4-d5 acid, L-Glutamic Acid-d5 |
| InChI Formula | InChI=1S/C5H9NO4/c6-3(5(9)10)1-2-4(7)8/h3H,1-2,6H2,(H,7,8)(H,9,10)/t3-/m0/s1/i1D2,2D2,3D |
| SMILES | [2H]C([2H])(C(=O)O)C([2H])([2H])[C@]([2H])(N)C(=O)O |
| IUPAC Name | (2S)-2-amino-2,3,3,4,4-pentadeuteriopentanedioic acid |
| Molecular Weight (g/mol) | 152.16 |
| Show More |