Learn More
L-gamma-Glutamyl-L-valine, TRC
Product Code. 30407966
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30407966 100 mg -
30392444 10 mg 10mg
30401678 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30407966 Supplier Toronto Research Chemicals Supplier No. TRCH253810100MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2746-34-1
Molecular Formula C10 H18 N2 O5
Molecular Weight (g/mol) 246.26
Synonym N-L-γ-Glutamyl-L-valine, N-L-γ-Glutamyl-(6CI,7CI) Valine, N-L-γ-Glutamyl-L-(8CI) Valine
IUPAC Name (2S)-2-amino-5-[[(1S)-1-carboxy-2-methylpropyl]amino]-5-oxopentanoic acid
SMILES CC(C)[C@H](NC(=O)CC[C@H](N)C(=O)O)C(=O)O
Specifications
InChI Formula InChI=1S/C10H18N2O5/c1-5(2)8(10(16)17)12-7(13)4-3-6(11)9(14)15/h5-6,8H,3-4,11H2,1-2H3,(H,12,13)(H,14,15)(H,16,17)/t6-,8-/m0/s1
Quantity 100 mg
Formula Weight 246.122
Percent Purity >95%
Shipping Condition Room Temperature
Chemical Name or Material L-gamma-Glutamyl-L-valine
Grade HPLC