missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ L-Ficolin Monoclonal Antibody (19)
GREENER_CHOICE

Product Code. 11534112
Change view
Click to view available options
Quantity:
400 μL
Unit Size:
200µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11534112 400 μL 200µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11534112 Supplier Invitrogen™ Supplier No. ABS0051902

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ABS 005-19-02 detects L-Ficolin from human samples. This antibody is specific towards L-Ficolin; no cross reactivity is seen with M-Ficolin (Ficolin-1) and H-Ficolin (Ficolin-3). ABS 005-19-02 has been successfully used in ELISA and WB procedures. NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL.

L-Ficolin (Ficolin-2) is an innate immunity pattern recognition molecule found in human serum. L-Ficolin binds to distinct pathogen-associated molecular patterns (PAMP), such as carbohydrates (GlcNAc), lipoteichoic acid and acetylated groups thereby facilitating phagocytosis and activation of complement through the lectin route using the same serine proteases as mannose-binding lectin (MBL) named MASP ((MBL)-associated serine protease).
TRUSTED_SUSTAINABILITY

Specifications

Antigen L-Ficolin
Applications ELISA, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 19
Concentration 1.02 mg/mL
Conjugate Unconjugated
Formulation PBS with 15mM sodium azide; pH 7.4
Gene FCN2
Gene Accession No. Q15485
Gene Alias 37 kDa elastin-binding protein; Collagen/fibrinogen domain-containing protein 2; EBP-37; FCN2; Fcnb; FCNL; ficolin (collagen/fibrinogen domain containing lectin) 2 (hucolin); ficolin 2; ficolin B; ficolin-2; Ficolin-B; ficolin-beta; Hucolin; L-ficolin; P35; RP11-263F14.2; Serum lectin p35
Gene Symbols FCN2
Host Species Mouse
Immunogen Purified recombinant protein corresponding to human L-Ficolin and full-length L-Ficolin (Ficolin-2).
Purification Method Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2220
Target Species Human
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.