missing translation for 'onlineSavingsMsg'
Learn More

L-Cysteinyl-L-cysteine TFA Salt (Technical Grade), TRC

Product Code. 30179161
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179161 10 mg 10mg
30179094 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30179161 Supplier Toronto Research Chemicals Supplier No. TRCC99999010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C6 H12 N2 O3 S2 . C2 H F3 O2
Molecular Weight (g/mol) 338.324
Synonym L-Cysteine, N-L-cysteinyl-TFA Salt, 140 TFA Salt: PN: EP2161028 PAGE TFA Salt: 10 claimed protein TFA Salt, 2416 TFA Salt: PN: WO2004035732 SEQID: 2418 claimed protein TFA Salt, 29: PN: US20040116339 SEQID: 38 claimed protein TFA Salt, 37: PN: WO2011146121 PAGE: 114 claimed sequence TFA Salt, Protein (human clone WO2004035732-SEQID-2418 fragment) TFA Salt
IUPAC Name (2R)-2-[[(2R)-2-amino-3-sulfanylpropanoyl]amino]-3-sulfanylpropanoic acid;2,2,2-trifluoroacetic acid
SMILES N[C@@H](CS)C(=O)N[C@@H](CS)C(=O)O.OC(=O)C(F)(F)F

Specifications

InChI Formula InChI=1S/C6H12N2O3S2.C2HF3O2/c7-3(1-12)5(9)8-4(2-13)6(10)11;3-2(4,5)1(6)7/h3-4,12-13H,1-2,7H2,(H,8,9)(H,10,11);(H,6,7)/t3-,4-;/m0./s1
Quantity 10 mg
Formula Weight 338.022
Shipping Condition Room Temperature
Chemical Name or Material L-Cysteinyl-L-cysteine TFA Salt (Technical Grade)
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.