missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Cysteine [Free Base], MP Biomedicals
Click to view available options
Quantity:
500 g
Specifications
Formula Weight | 121.2 |
Grade | Cell Culture Reagent |
Quantity | 500 g |
Physical Form | Crystalline |
Chemical Name or Material | L-Cysteine [Free Base] |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'