Learn More
L-Amoxicillin, TRC
Specifications
Specifications
| Chemical Name or Material | L-Amoxicillin |
| CAS | 26889-93-0 |
| Molecular Formula | C16 H19 N3 O5 S |
| Quantity | 100 mg |
| Synonym | L-Amoxicillin, 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 6-[[(2S)-amino(4-hydroxyphenyl)acetyl]amino]-3,3-dimethyl-7-oxo-, (2S,5R,6R)- (9CI), 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 6-[2-amino-2-(p-hydroxyphenyl)acetamido]-3,3-dimethyl-7-oxo-, stereoisomer (8CI), 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 6-[[amino(4-hydroxyphenyl)acetyl]amino]-3,3-dimethyl-7-oxo-, [2S-[2α,5α,6β(R*)]]-, (2S,5R,6R)-6-[[(2S)-2-Amino-2-(4-hydroxyphenyl)acetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, Amoxicillin Trihydrate Imp. B (EP) |
| InChI Formula | InChI=1S/C16H19N3O5S/c1-16(2)11(15(23)24)19-13(22)10(14(19)25-16)18-12(21)9(17)7-3-5-8(20)6-4-7/h3-6,9-11,14,20H,17H2,1-2H3,(H,18,21)(H,23,24)/t9-,10+,11-,14+/m0/s1 |
| SMILES | CC1(C)S[C@@H]2[C@H](NC(=O)[C@@H](N)c3ccc(O)cc3)C(=O)N2[C@H]1C(=O)O |
| IUPAC Name | (2S,5R,6R)-6-[[(2S)-2-amino-2-(4-hydroxyphenyl)acetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid |
| Molecular Weight (g/mol) | 365.4 |
| Formula Weight | 365.1045 |