missing translation for 'onlineSavingsMsg'
Learn More

L-Alanine tert-butyl ester hydrochloride, 98%, Thermo Scientific Chemicals

Product Code. p-7039875
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15405648 1 g 1g
15415648 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15405648 Supplier Thermo Scientific Alfa Aesar Supplier No. H59359.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Soluble in methanol.

Notes
Hygroscopic. Keep away from heat and humidity. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13404-22-3
Molecular Formula C7H16ClNO2
Molecular Weight (g/mol) 181.66
MDL Number MFCD00035524
InChI Key WIQIWPPQGWGVHD-JEDNCBNOSA-N
Synonym h-ala-otbu.hcl, tert-butyl l-alaninate hydrochloride, l-alanine tert-butyl ester hydrochloride, s-tert-butyl 2-aminopropanoate hydrochloride, tert-butyl alaninate hydrochloride, h-ala-otbu inverted exclamation mark currencyhcl, tert-butyl 2s-2-aminopropanoate hydrochloride, l-alanine t-butyl ester hydrochloride, alanine, 1,1-dimethylethyl ester, hydrochloride, l-alanine, 1,1-dimethylethyl ester, hydrochloride
PubChem CID 11275390
SMILES Cl.C[C@H](N)C(=O)OC(C)(C)C

Specifications

Melting Point 168°C to 175°C
Beilstein 3562140
Solubility Information Soluble in methanol.
IUPAC Name tert-butyl 2-aminopropanoate hydrochloride
Quantity 1 g
Formula Weight 181.66
Optical Rotation +1.4° (c=2 in Ethanol)
Sensitivity Hygroscopic
Percent Purity 98%
Chemical Name or Material L-Alanine tert-butyl ester hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.