Learn More
MedChemExpress L-(+)-Abrine
Specifications
Specifications
| Content And Storage | Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month |
| Molecular Weight (g/mol) | 218.25 g/mol |
| Shipping Condition | Room temperature |
| For Use With (Application) | Neuroscience-Neuromodulation |
| CAS | 526-31-8 |
| Product Type | Natural Products |
| Purity | 99.76% |
| Solubility | H2O : 2 mg/mL (9.16 mM∣ultrasonic and adjust pH to 10 with NaOH) |
| Synonym | L-Abrine∣ L-N-Methyltryptophan∣ N-α-Methyl-L-tryptophan |
| Quantity | 100 mg |