Learn More
L 888607 Racemate, MedChemExpress
Product Code. 16487137
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. 16487137
Brand: Medchem Express HY111271A
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress L 888607 Racemate is a selective prostaglandin D2 receptor subtype 1 (DP1) antagonist, with Kis of 132 nM and 17 nM for DP1 and thromboxane A2 receptor (TP), respectively.
L 888607 Racemate is a selective prostaglandin D2 receptor subtype 1 (DP1) antagonist, with Kis of 132 nM and 17 nM for DP1 and thromboxane A2 receptor (TP), respectively.
Specifications
Chemical Name or Material L 888607 Racemate
CAS 1030017-51-6
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C19H15ClFNO2S
Quantity 5 mg
Solubility Information DMSO : 6 mg/mL (15.96 mM; Need ultrasonic)
SMILES O=C(O)CC1CCN2C1=C(SC3=CC=C(Cl)C=C3)C4=C2C=C(F)C=C4
Molecular Weight (g/mol) 375.84
Formula Weight 375.84
Percent Purity 99.44%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation