missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-685458, MedChemExpress
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
Description
L-685458 inhibits amyloid β-protein precursor γ-secretase activity with IC50 of 17 nM, shows greater than 50-100-fold selectivity over other aspartyl proteases tested. L685458 inhibits γ-secretase-mediated cleavage of APP-C99 and Notch-100 with IC50s of 301.3 nM and 351.3 nM, respectively. L-685458 can be used for the research of alzheimer’s disease (AD) and cancers.
Specifications
Specifications
| Chemical Name or Material | L-685458 |
| Physical Form | Solid |
| CAS | 292632-98-5 |
| Color | White |
| Quantity | 5 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C39H52N4O6 |
| For Use With (Application) | Cancer-programmed cell death |
| Synonym | L-685,458 |
| Solubility Information | DMSO : 83.3 mg/mL (123.80 mM; Need ultrasonic and warming) |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction