missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KSV Cyclin D Antibody (94B), Alexa Fluor™ 647, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NB100-2621AF647
This item is not returnable.
View return policy
Description
KSV Cyclin D Monoclonal specifically detects KSV Cyclin D in Virus samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| KSV Cyclin D | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Virus | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| 94B | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| Kaposi's Sarcoma Virus Type D Cyclin | |
| Recombinant fusion protein produced from a clone from KSV. | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication, Core ESC Like Genes, mTOR Pathway, Stem Cell Markers, Wnt Signaling Pathway | |
| 595 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction