missing translation for 'onlineSavingsMsg'
Learn More

Elma™ KS Rinsing Solution

Product Code. 11751369
Change view
Click to view available options
Volume (Metric):
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
11751369 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11751369 Supplier Elma™ Supplier No. 5820600000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

KS Rinsing Solution is for the temporary corrosion protection used as additive to the rinsing or cleaning bath.

  • Elma KS Rinsing Solution is a liquid, mildly alkaline concentrate for the temporary corrosion protection used as additive to the rinsing or cleaning bath.
  • Corrosion-sensitive metals such as steel, gray cast iron and hard metals are covered with a protective coating on a molecular level which protects temporarily from corrosion.
  • Subsequent coating (PVD, CVD) or hardening processes (plasma- and carbonitriding) are not impeded.

Specifications

Form Liquid
pH 9 to 9.5
Concentration 0.05 to 0.5%
Container Type Bottle
Content And Storage -5°C to 30°C
Volume (Metric) 1 L
compliance-icons
Product Identifier
  • elma korrosionsschutzzusatz (elma-KS)
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P102-Keep out of reach of children.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
Supplemental information
  • MIXTURE LIST-Contain: 2-aminoethoxy ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.