missing translation for 'onlineSavingsMsg'
Learn More

KRN-633, MedChemExpress

Product Code. p-200099417
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16491207 50 mg 50mg
16487697 5 mg 5mg
16475736 100 mg 100mg
16414968 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16491207 Supplier Medchem Express Supplier No. HY12060

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress KRN-633 is a potent VEGFR inhibitor with IC50s of 170, 160 and 125 nM for VEGFR1, VEGFR2 and VEGFR3, respectively.

KRN-633 is a potent VEGFR inhibitor with IC50s of 170, 160 and 125 nM for VEGFR1, VEGFR2 and VEGFR3, respectively.

Specifications

Chemical Name or Material KRN-633
CAS 286370-15-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C20H21ClN4O4
Quantity 50 mg
Solubility Information DMSO : 25 mg/mL (59.97 mM; Need ultrasonic)
SMILES ClC1=CC(OC2=NC=NC3=CC(OC)=C(C=C32)OC)=CC=C1NC(NCCC)=O
Molecular Weight (g/mol) 416.86
Formula Weight 416.86
Percent Purity 99.37%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.