Learn More
KR-33493, MedChemExpress
Description
KR-33493 is a potent inhibitor of Fas-mediated cell death (FAF1).
Specifications
Specifications
| Chemical Name or Material | KR-33493 |
| CAS | 1021497-97-1 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C20H18BrN3O3S |
| Quantity | 50 mg |
| Solubility Information | DMSO : ≥ 31 mg/mL (67.34 mM) |
| SMILES | OC(C1=NN(CCC2=CC=CC=C2)C=C1NC(CSC3=CC=C(Br)C=C3)=O)=O |
| Molecular Weight (g/mol) | 460.34 |
| Formula Weight | 460.34 |
| Show More |