missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ Kontes™ Polypropylene Screw Thread with Open Top

Brand:  DWK Life Sciences 73806A-20400

Additional Details : Weight : 0.10000kg

Product Code. 10798404

  • 207.00 € / Pack of 1000
Estimated Shipment: 25-07-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

PTFE/Silicone
1.2 cm
Polypropylene
20-400
0.472 in.
Black
Autoclavable
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions