missing translation for 'onlineSavingsMsg'
Learn More

Bode Chemie™ Kohrsolin™ Surface Disinfectant

Product Code. p-8000878
Click to view available options
pH:
3.2
4
Unit Size:
5 L
Pack of 250
This item is not returnable. View return policy

Product Code. 10415963

Brand: Bode Chemie™ 973623

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Use Bode Chemie™ Kohrsolin™ Surface Disinfectant for the prophylactic cleaning wipe disinfection of all surfaces in areas relevant to hygiene. It is also suitable for targeted disinfection measures requiring an extended spectrum of activity (e.g. virucidal activity).

  • Certifications/Compliance: CE, MDD, VAH
  • Shelf Life: 12Months
  • Composition: Liquid or sachet

Specifications

Product Type Surface Disinfectant Cleaner
Form Liquid
pH 4
Appearance Light Yellow
Certifications/Compliance CE, MDD, VAH
Composition Active ingredients: (ethylenedioxy) dimethanol 141mg/g, Glutaral 50mg/g, Didecyldimethylammonium chl
Container Type Canister
Volume (Metric) 5000 mL
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
  • Germ cell mutagenicity Category 2
  • Carcinogenicity Category 1
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.
  • H302 + H332-Harmful if swallowed or if inhaled.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501-Dispose of contents/container to .
  • P310-Immediately call a POISON CENTER/doctor/.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.