missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bode Chemie™ Kohrsolin™ Surface Disinfectant
Click to view available options
pH:
3.2
4
Unit Size:
5 L
Pack of 250
Description
- Certifications/Compliance: CE, MDD, VAH
- Shelf Life: 12Months
- Composition: Liquid or sachet
Specifications
Specifications
| Product Type | Surface Disinfectant Cleaner |
| Form | Liquid |
| pH | 3.2 |
| Appearance | Light Yellow |
| Certifications/Compliance | CE, MDD, VAH |
| Composition | Active ingredients: (ethylenedioxy) dimethanol 141mg/g, Glutaral 50mg/g, Didecyldimethylammonium chl |
| Container Type | Canister |
| Volume (Metric) | 5000 mL |
Safety and Handling
Signal Word
- Danger
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Respiratory sensitiser Category 1
- Serious eye damage/eye irritation Category 1
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 2
- H302 + H332-Harmful if swallowed or if inhaled.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H335-May cause respiratory irritation.
- H400-Very toxic to aquatic life.
- H411-Toxic to aquatic life with long lasting effects.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P310-Immediately call a POISON CENTER/doctor/.
- P501-Dispose of contents/container to .
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction