Learn More
Bode Chemie™ Kohrsolin FF Surface Disinfectant Cleaner
Product Code. 10731391
Change view
Click to view available options
Volume (Metric):
40 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
10731391 40 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10731391 Supplier Bode Chemie™ Supplier No. 9804283
  • ONLINE EXCLUSIVE PRICE
    129.00 €
    62.49 € / Pack of 1
    Save 66.51 €
    51% Off
 Restricted Item
Estimated Shipment: 13-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Product Type Surface Disinfectant Cleaner
Certifications/Compliance VAH, MDD
Composition Active ingredients: Glutaral 50mg /g, Benzyl-C12-18-alkyldimethyl-ammonium chlorides 30mg /g, didecy
Container Type Sachet
Volume (Metric) 40 mL