missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KNT1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5119465
This item is not returnable.
View return policy
Description
KNT1 Polyclonal Antibody for Western Blot, IHC (P)
KNT1 (T-Kinninogen 1) is the acute phase kininogen of the rat. KNT1 could be a healing protein because of its properties as a cysteine protease inhibitor. Evidence has been presented that mRNA of T-kininogen synthesis may be a function of interleukin 6 production. In rats, four types of kininogens are produced, two of which are classical high and low molecular weight kininogens and two of which are low molecular weight-like kininogens, designated T-kininogen 1 and T-kininogen 2. T-kininogen 1 and T-kininogen 2 are 430 amino acid secreted rat proteins that each contain three cystatin domains and have nearly identical functions. Existing in plasma, both T-kininogen 1 and T-kininogen 2 are glycoproteins that act as thiol protease inhibitors and also play a role in blood coagulation, specifically by helping to optimally position blood coagulation factors.
Specifications
| KNT1 | |
| Polyclonal | |
| Unconjugated | |
| Kng1 | |
| T-Kinninogen 1 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 288001 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P08934 | |
| Kng1 | |
| Recombinant protein T-Kininogen 1. The antigen corresponds to amino acid range 378-430 of the target protein. | |
| 200 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction