Learn More
Carl Roth™ Know Label
Click to view available options
Diameter (Metric):
12 mm
Specifications
Specifications
| Product Type | Label |
| For Use With (Application) | Without drying out and falling off sticking on any plastic, z. B. during steam sterilization, in boiling water baths, in refrigerators, in agitation and shakers, centrifuges etc. |
| Temperature Range (Metric) | -96 to 80°C |
| Diameter (Metric) | 12 mm |