Learn More
Carl Roth™ Know Label
Product Code. p-8006270
Encompass
Click to view available options
Diameter (Metric):
12 mm
Product Code. 10761333
Brand: Carl Roth™ N120.1
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.
Request a Price for this item here.
Specifications
Product Type Label
For Use With (Application) Without drying out and falling off sticking on any plastic, z. B. during steam sterilization, in boiling water baths, in refrigerators, in agitation and shakers, centrifuges etc.
Temperature Range (Metric) -96 to 80°C
Diameter (Metric) 12 mm