Learn More
KLHL2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14449-1-AP-20UL
This item is not returnable.
View return policy
Description
KLHL2 (kelch-like protein 2), also known as actin-binding protein Mayven, is a 593 amino acid actin-binding protein. Highly expressed in brain, KLHL2 is thought to play a role in the organization of the actin cytoskeleton in brain cells.Specifications
| KLHL2 | |
| Polyclonal | |
| Unconjugated | |
| KLHL2 | |
| ABP KELCH, Actin binding protein Mayven, Kelch like protein 2, KLHL2, MAV, MAYVEN | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11275, 290692, 77113 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Western Blot, Immunocytochemistry | |
| 0.22 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| F1LZF0, O95198, Q8JZP3 | |
| Klhl2 | |
| KLHL2 Fusion Protein Ag5950 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |