Learn More
KLHL12 Monoclonal Antibody (OTI4E1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA803209S
Encompass_Preferred
 View more versions of this product
Product Code. 17552153
  • 242.00 € / 30µL
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
KLHL12 Monoclonal antibody specifically detects KLHL12 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot
Specifications
Specifications
KLHL12
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P33897
Mouse
Affinity chromatography
RUO
215
-20° C, Avoid Freeze/Thaw Cycles
IgG2b
Immunohistochemistry (Paraffin), Western Blot
OTI4E1
Unconjugated
ABCD1
C3IP1; CUL3-interacting protein 1; DKIR; DKIR homolog; FLJ27152; hDKIR; kelch like family member 12; kelch-like 12; kelch-like family member 12; kelch-like protein 12; kelch-like protein C3IP1; Klhl12; si:dkeyp-53e12.5; Unknown (protein for MGC:134560); wu:fi60g03; zgc:92570
Human recombinant protein fragment corresponding to amino acids 508-745 of human ABCD1 produced in E.coli.
30 μL