Learn More
KLHL12 Monoclonal Antibody (OTI4E1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA803209
Description
Serves as a substrate-specific adapter for the CUL3-based ubiquitin-protein E3 ligase complex. Negatively regulates the Wnt signaling pathway via the targeted ubiquitination and subsequent proteolysis of DVL3.Specifications
| KLHL12 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q53G59 | |
| KLHL12 | |
| Human recombit protein fragment corresponding to amino acids 177-405 of human KLHL12 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI4E1 | |
| Unconjugated | |
| KLHL12 | |
| C3IP1, DKIR | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 59349 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.