Learn More
Thermo Scientific™ Klenow Fragment
Klenow Fragment is the Large Fragment of DNA Polymerase exhibiting 5'→3' polymerase, 3'→5' exonuclease (proofreading) activities, but lacking 5'→3' exonuclease activity.
Brand:  Thermo Scientific™ EP0051-RMD123
Additional Details : Weight : 0.00900kg
Product Code. 15619454
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Highlights
  • Incorporates modified nucleotides (e.g., Cy3-, Cy5-, aminoallyl-, biotin-, digoxigenin- and fluorescently-labeled nucleotides)
  • Active in restriction enzyme, PCR, RT, and T4 DNA Ligase buffers
Applications
  • DNA blunting by fill-in 3'-overhangs
  • Random-primed DNA labeling
  • Labeling by fill-in 5'-overhangs of dsDNA
  • DNA sequencing by the Sanger method
  • Site-specific mutagenesis of DNA with synthetic oligonucleotides
  • Second strand synthesis of cDNA