missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
This gene encodes a type I transmembrane protein and member of the immunoglobulin superfamily of cell adhesion molecules. The encoded protein localizes to adherens junctions in pancreatic beta cells and regulates insulin secretion. Autoantibodies against the encoded protein have been detected in serum from patients with type 1 diabetes. This gene may also play a role in glomerular development and decreased expression of this gene has been observed in human glomerular diseases. This gene and the related opposite-strand gene nephrin are regulated by a bidirectional promoter.
Specifications
Specifications
| Antigen | KIRREL2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.41 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| Gene | KIRREL2 |
| Gene Accession No. | Q6UWL6, Q7TSU7 |
| Gene Alias | FILTRIN, Kin of IRRE like protein 2, KIRREL2, NEPH3, Nephrin like protein 3, NLG1 |
| Gene Symbols | Kirrel2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?