missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ KIR2DL1/KIR2DS5 Antibody (Innate patent anti-KIR2DL) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28027-1mg
This item is not returnable.
View return policy
Description
KIR2DL1/KIR2DS5 Monoclonal antibody specifically detects KIR2DL1/KIR2DS5 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| KIR2DL1/KIR2DS5 | |
| Monoclonal | |
| Unconjugated | |
| P43626 | |
| Human | |
| Protein A purified | |
| RUO | |
| 3802 | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Functional Assay | |
| Innate patent anti-KIR2DL | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| CD158a | |
| KIR2DL1/ CD158a | |
| 1 mg | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction