missing translation for 'onlineSavingsMsg'
Learn More

KIN1408, MedChemExpress

Product Code. 16437988
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16437988 10 mg 10mg
16490448 5 mg 5mg
16457766 100 mg 100mg
16403917 50 mg 50mg
16401909 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16437988 Supplier Medchem Express Supplier No. HY19961

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress KIN1408 is an agonist of the RIG-1-like receptor (RLR) pathway and exhibits a broad-spectrum antiviral activity. KIN1408 exhibits activity against HCV, influenza A, dengue virus 2, Ebola, Nipah, and Lassa viruses.

KIN1408 is an agonist of the RIG-1-like receptor (RLR) pathway and exhibits a broad-spectrum antiviral activity. KIN1408 exhibits activity against HCV, influenza A, dengue virus 2, Ebola, Nipah, and Lassa viruses.

Specifications

Chemical Name or Material KIN1408
Physical Form Solid
CAS 1903800-11-2
Color Light Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C25H19F2N3O3S
For Use With (Application) COVID-19-anti-virus
Solubility Information DMSO : ≥ 39 mg/mL (81.33 mM)
SMILES COC1=CC=C(SC(NC(C2=CC=C(OC(F)F)C=C2)C3=C(O)C(N=CC=C4)=C4C=C3)=N5)C5=C1
Molecular Weight (g/mol) 479.5
Formula Weight 479.5
Percent Purity 98.78%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.