Learn More
Kimtech™ G3 NxT Nitrile Gloves
GREENER_CHOICE
Product Code. 17807045
Click to view available options
Size:
Large
Large+
Medium
Small
X-Large
X-Small
Unit Size:
Pack of 1000
Product Code. 17807045
Brand: Kimtech™ 6299440
Only null left
Add to basket
Add to basket
Nitrile gloves provide seamless protection and are suitable for a range of cleanroom environments.
  • Rigorously tested to ensure regulatory compliance, making them suitable for ISO Class 3 or higher cleanroom environments
  • Synthetic nitrile polymer material is designed for fit and reliability, with textured fingertips for improved grip and excellent water tightness that results in a low risk of pinholes
  • Ambidextrous and incorporate a beaded cuff for added strength and ease of donning, so the wearer can simply grab and go without any fear of ripping the material
  • Latex, silicone and powder-free
  • The absence of natural rubber latex reduces the risk of TYPE 1 glove-associated reactions, protecting the wearer as well as the application
  • Designated as PPE Cat III according to (EU) Regulation 2016/425 and to the Regulation 2016/425 as brought into UK law and amended
  • Gloves are provided packaged in cleanroom-compatible polyethylene so as to be easily integrated into your processes
Specifications
Certifications/Compliance (Europe) PPE Cat III according to Regulation (EU) 2016/425 and to the Regulation 2016/425 as brought into UK law and amended
Chlorinated Yes
Cleanroom Class Class 1 (ISO 3)
Accelerator Free No
AQL 1.5
Color White
Disposable Single-use
Exterior Finish Fingertip Textured
Finger Thickness (English) 6.3 mil
Finger Thickness (Metric) 0.16 mm
Hand Ambidextrous
Latex Free Yes
Material Nitrile
Palm Thickness (English) 5 mil
Palm Thickness (Metric) 0.13 mm
Powdered Powder Free
Size X-Large
Sterility Non-sterile
Thickness (English) 0.004 in.
Thickness (Metric) 0.10 mm
Manufacturing/QMS Audit Standards ISO 9001 and ISO 13485
Model 62994-40
Tensile Strength 18 MPa
Length (English) 12 in.
Length (Metric) 30.5 cm