Learn More
DWK Life Sciences Kimble Chase; Kimble™ Kontes™ ULTRA-WARE™ Filtration Adapter
Product Code. 10062622
Click to view available options
:
Filtration adapter assembly for 32mm
Product Code. 10062622
Brand: DWK Life Sciences 1799504532
Please to purchase this item. Need a web account? Register with us today!
This adapter assembly is designed to allow connection of a 47mm or 90mm filtration support base with a straight drip tip to a bottle or reservoir with a GL 45 thread.
  • Filtrate is collected directly into a media-type bottle when using a standard membrane filtration assembly
  • All components are steam-autoclavable
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
TRUSTED_SUSTAINABILITY
Specifications
Type Filtration Adapter Assembly
Diameter (Metric) Inside 32 mm