Learn More
DWK Life Sciences Kimble™ Kontes™ Tube For Potter-Elvehjem Tissue Grinder
Product Code. 10355402
Change view
Click to view available options
Size:
19
20
21
23
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
10355402 20 Each
10633823 23 Each
10315362 19 Each
10613633 21 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10355402 Supplier DWK Life Sciences Supplier No. 8855120020
Only null left
Add to basket
Add to basket
A graduated tube with markings fused onto the glass. Range of tube sizes: Size 20 (4mL), Size 21 (10mL) and Size 22 (55mL).
  • Graduations are fused onto the glass
  • Clearance between pestle and tube
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
TRUSTED_SUSTAINABILITY
Specifications
Size 20
Product Type Tissue Grinder Tube
For Use With (Equipment) Potter-Elvehjem tissue grinder