Learn More
DWK Life Sciences Kimble™ Duall™ All-Glass Tissue Grinder Accessory, Grinding Tube
Description
Improves grinding efficiency
- Tubes sizes from 21 to 25
- Available with or without coating
Specifications
Specifications
| Size | 21 |
| Product Type | Tissue Grinder Tube |
| For Use With (Application) | Improves grinding efficiency |
| Includes | Without plastic safety coating |
| For Use With (Equipment) | Duall™ all Glass tissue grinders |