Learn More
DWK Life Sciences Kimble™ Kontes™ Chromatography Flow Control Adapter with 24/40 ST Joint
Product Code. 15883002
Change view
Click to view available options
Packaging Quantity:
1 Ea.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Packaging Quantity unitSize
15883002 1 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15883002 Supplier DWK Life Sciences Supplier No. 4205752440
Only null left
Add to basket
Add to basket
The flow control adapter features Kimble™ Kontes™ Hi-Vac™ PTFE plug designed with a precision external thread for fine control.
  • The flow control adapter features Kontes Hi-Vac™ PTFE plug designed with a precision external thread for fine control
  • The accurately controlled valve seat and the extended tip on the PTFE plug allow semi-needle valve control for liquids
  • The FKM o-ring is shielded from direct exposure to liquids within the system by PTFE ring seals
TRUSTED_SUSTAINABILITY
Specifications
Type Flow Control Adapter
Packaging Quantity 1 Ea.
Includes Joint