Learn More
DWK Life Sciences Kimble™ Kontes™ Chromatography Flow Control Adapter with 24/40 ST Joint
Description
- The flow control adapter features Kontes Hi-Vac™ PTFE plug designed with a precision external thread for fine control
- The accurately controlled valve seat and the extended tip on the PTFE plug allow semi-needle valve control for liquids
- The FKM o-ring is shielded from direct exposure to liquids within the system by PTFE ring seals
Specifications
Specifications
| Type | Flow Control Adapter |
| Packaging Quantity | 1 Ea. |
| Includes | Joint |