Learn More
DWK Life Sciences Kimble™ Kontes™ Clamps
Product Code. 11883123
Click to view available options
Type:
Aluminum Clamp
Support Base
Unit Size:
Each
Product Code. 11883123
Brand: DWK Life Sciences 9538410090
Only null left
Add to basket
Add to basket
For 90mm filter
  • Component part for all-glass ultra-ware microfiltration apparatus with stainless steel support
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) All-glass ultra-ware microfiltration apparatus with stainless steel support
Material Glass
Standard Taper Joint 40/35
Type Support Base
Diameter (Metric) Inside 90 mm