Learn More
DWK Life Sciences Kimble™ Aluminum Seal Caps
Click to view available options
Color:
Natural
Red
No. per Case:
100/Cs.
1000/Cs.
Unit Size:
Pack of 100
Pack of 1000
Description
- Moderate to good resealability
- One piece
- PTFE-faced red rubber septa
- For general GC applications
- Moderate resealability
- Not recommended for multiple injections
- PTFE-faced natural rubber septa
- Good resealability
- Multiple injections
Specifications
Specifications
| Color | Natural |
| For Use With (Application) | Multiple injection |
| Material | Aluminum |
| No. per Case | 100/Cs. |
| Liner Material | PTFE/Natural Silicone Rubber |
| Neck Finish | 11 |