Learn More
DWK Life Sciences Kimble™ KIMAX™ Long, Straight-Neck Flask with Capacity Scale
Product Code. p-122297
Click to view available options
Capacity (Metric):
250 mL
Unit Size:
Pack of 24
Product Code. 10389924
Brand: DWK Life Sciences 25615250
Only null left
Add to basket
Add to basket
Neck accepts metal or plastic cap or cotton plug
  • Thick-walled tubing neck and plain fire-polished finish sealed to an Erlenmeyer-shaped body
  • Graduated to capacity
  • Durable white ceramic enamel scale
Compliance
Designed from ASTM™ E 1404, Type IV, Class 1 requirements
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) Outer Neck 3.8 cm
Capacity (Metric) 250 mL
Material Borosilicate Glass
For Use With (Application) Metal cap, Plastic closure, Foam plug, or Cotton plug for Cell Culture applications
Graduation Range 50 to 225 mL
Graduations 25 mL
Height (English) 5.5 in.
Height (Metric) 14.2 cm
Neck Style Plain fire-polished
Width (English) 3.2 in.
Width (Metric) 8.2 cm
Autoclavable Autoclavable
Closure Size 38 mm