Learn More
DWK Life Sciences Kimble™ KIMAX™ GL 45 Media/Storage Bottles
Product Code. p-3206618
Click to view available options
Capacity (Metric):
500 mL
1000 mL
Graduations:
100 to 400mL
100 to 900mL
Unit Size:
Pack of 10
Product Code. 11780209
Brand: DWK Life Sciences 14395500
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Perfect for mixing, storing or transporting culture media, chemicals or solvents
  • Autoclavable Type 1 borosilicate glass
  • Permanent white-enamel graduations and marking spots
  • Approx. 30mm I.D. GL 45 screw-thread opening
  • Linerless blue polypropylene cap autoclavable to 140°C (284°F) and replaceable, drip-free, clear polypropylene pour ring included
Order Info
Cap and ring also sold separately; contact your Fisher Scientific™ Customer Service Representative.
TRUSTED_SUSTAINABILITY
Specifications
Capacity 17 oz., 500 mL
Capacity (Metric) 500 mL
Capacity (English) 17 oz.
Color White
Diameter (English) 3.3 in.
Diameter (English) Inner Neck 1.18 in.
Diameter (Metric) Inner Neck 30 mm
Includes Clear drip-free Polypropylene pouring ring
Material Borosilicate Glass (Type I)
Closure Color Blue
Graduations 100 to 400mL
Closure Included Yes
Graduated Yes
Shape Round
Height (Metric) 176 mm
Product Type Media Bottle
Height (English) 6.9 in.
Closure Material Polypropylene
Closure Size GL 45
Diameter (Metric) 86 mm
Closure Type Screw Cap
Autoclavable Autoclavable