missing translation for 'onlineSavingsMsg'
Learn More

KIF7 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Product Code. 18451426 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18451426 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18451426 Supplier Novus Biologicals Supplier No. NBP146838JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

KIF7 Polyclonal antibody specifically detects KIF7 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIF7
Applications Western Blot
Classification Polyclonal
Conjugate Janelia Fluor 646
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias EQYK340, kinesin family member 7, kinesin-like protein KIF7, MGC120653, MGC138476, MGC138478, UNQ340
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 1225 and 1275 of human kinesin family member 7 using the numbering given in entry NP_940927.2 (GeneID 374654).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cytoskeleton Markers, Signal Transduction, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 374654
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.