missing translation for 'onlineSavingsMsg'
Learn More

KIF3A Antibody, Allophycocyanin, Novus Biologicals™

Product Code. 30034648 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30034648 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30034648 Supplier Novus Biologicals Supplier No. NBP146837APC

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

KIF3A Polyclonal antibody specifically detects KIF3A in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIF3A
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate APC
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias KIF3, kinesin family member 3A, kinesin family protein 3A, kinesin-like protein KIF3A, Microtubule plus end-directed kinesin motor 3A
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 652 and 702 of human kinesin family member 3A using the numbering given in entry AAC72294.1 (GeneID 11127).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cytoskeleton Markers, Immunology, Neuroscience, Signal Transduction, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 11127
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.