missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ KIAA0391 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16520722
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16520722 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16520722 Supplier Invitrogen™ Supplier No. PA597892

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

KIAA0391 Polyclonal Antibody for Western Blot, IHC (P), ELISA

KIAA0391, also known as MRPP3 (mitochondrial ribonuclease P protein 3), is a 583 amino acid protein that is located in the mitochondria. KIAA0391 functions in tRNA maturation and is a component of mitochondrial ribonuclease P which cleaves tRNA at the 5'-end. Mitochondrial ribonuclease P is an enzyme composed of RG9MTD1, MRPP22 and KIAA0391 subunits. Due to alternative splicing events, KIAA0391 exists as four isoforms. The gene encoding KIAA0391 maps to human chromosome 14, which contains about 700 genes and makes up about 3. 5% of human cellular DNA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIAA0391
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.23 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PRORP
Gene Accession No. O15091
Gene Alias 1110008L16Rik; KIAA0391; Mitochondrial ribonuclease P catalytic subunit; Mitochondrial ribonuclease P protein 3; mitochondrial RNase P protein 3; mitochondrial RNase P subunit 3; mKIAA0391; MRPP3; Prorp; Protein only RNase P catalytic subunit; proteinaceous RNase P; RGD1305089; RIKEN cDNA 1110008L16 gene; similar to 1110008L16Rik protein
Gene Symbols PRORP
Host Species Rabbit
Immunogen Recombinant Human Mitochondrial ribonuclease P catalytic subunit protein (314-583AA).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9692
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.