Learn More
Ki8751, MedChemExpress
Product Code. 16494938
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Product Code. 16494938
Brand: Medchem Express HY12038
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Ki8751 is a potent VEGFR2 inhibitor with an IC50 of 0.9 nM.
Ki8751 is a potent VEGFR2 inhibitor with an IC50 of 0.9 nM.
Specifications
Chemical Name or Material Ki8751
CAS 228559-41-9
Color Off-White
Purity Grade Notes Research
Molecular Formula C24H18F3N3O4
Quantity 10 mg
Solubility Information DMSO : ≥ 92 mg/mL (195.99 mM)
SMILES COC1=CC2=C(C=CN=C2C=C1OC)OC3=CC=C(C(F)=C3)NC(NC4=CC=C(C=C4F)F)=O
Molecular Weight (g/mol) 469.41
Formula Weight 469.41
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease