Learn More
Tocris Bioscience™ KH-4-43
Description
KH-4-43 is a selective CUL4 inhibitor (Kd = 83 nM). KH-4-43 inhibits the ubiquitination of CK1α by CRL4CRBN in vitro. Treatment of cells with KH-4-43 causes accumulation of the E3 CRL4 substrate CDT1. KH-4-43 suppresses the growth of human tumor xenografts in mice.
Specifications
Specifications
| Chemical Name or Material | 3-[1-(4-Chlorophenyl)-1H-pyrazol-4-yl]-7,8-dihydroxy-2-(trifluoromethyl)-4H-1-benzopyran-4-one |
| CAS | 2813310-07-3 |
| Quantity | 5 mg |
| Target | Ubiquitin Ligase (E3) Inhibitors |
| Molecular Formula | C19H10ClF3N2O4 |
| Purity | 0.98 |