Learn More
Lonza™ KGM™ Gold Keratinocyte Growth Basal Medium
Product Code. 11605090
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11605090 500 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11605090 Supplier Lonza™ Supplier No. 192151
Please to purchase this item. Need a web account? Register with us today!
Separately sold keratinocyte basal medium for high and low density NHEK-Normal Human Epidermal Keratinocyte cultures.
  • Keratinocyte basal medium sold separately from combination media kit.
  • Can be used with 1 kit of 192152 (keratinocyte cell growth medium SingleQuots).
  • Designed to support both clonal growth and high density keratinocyte proliferation.
  • Supports superior growth and morphology of keratinocytes.
TRUSTED_SUSTAINABILITY
Specifications
Cell Type Keratinocyte Cells
Quantity 500 mL
Format 500 mL Bottle
Content And Storage KBM Gold Basal Medium