missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ KCTD5 Monoclonal Antibody (OTI3C8)
GREENER_CHOICE

Product Code. 15944674
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15944674 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15944674 Supplier Invitrogen™ Supplier No. MA525069

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

KCTD5 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow

KCTD5 is involved in interaction with CUL3 suggests that it may act as a substrate adapter in some E3 ligase complex. Does not affect the function of Kv channel Kv2.1/KCNB1, Kv1.2/KCNA2, Kv4.2/KCND2 and Kv3.4/KCNC4.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCTD5
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI3C8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene KCTD5
Gene Accession No. Q9NXV2
Gene Alias 2610030N08Rik; BTB/POZ domain-containing protein KCTD5; KCTD5; mKIAA0176; potassium channel tetramerisation domain containing 5; potassium channel tetramerization domain containing 5
Gene Symbols KCTD5
Host Species Mouse
Immunogen Full length human recombinant protein of KCTD5 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54442
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.